Learn More
Invitrogen™ S100A8 Human ELISA Kit
Descripción
The Human S100A8 ELISA quantitates Hu S100A8 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu S100A8. Principle of the method The Human S100A8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Especificaciones
Especificaciones
| Número de acceso | P05109 |
| Intervalo del ensayo | 10.24-2500 pg/mL |
| Sensibilidad del ensayo | 10 pg/mL |
| Intervalo de calibración | 10pg/mL to 2500pg/mL |
| ensayo | ELISA sandwich de fase sólida |
| Conjugado | Biotina |
| Tipo de producto | ELISA Kit |
| Familia de proteínas | Chemokine & Receptors |
| Tipo de muestra | Plasma, Serum, Supernatant |
| Especificidad | This ELISA pair antibody detects human S-100A8. Other species not determined. |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.