Learn More
ACROBiosystems Human Siglec-3 / CD33 Protein, Fc Tag
350.80€ - 2153.18€
Especificaciones
Número de acceso | AAH28152.1 |
---|---|
Peso molecular | 52.9 kDa |
Fuente | Human Siglec-3, Fc Tag (CD3-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). |
Inmunógeno | Asp 18 - His 259 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15876167
|
ACROBiosystems
CD3-H5257-100UG |
100 μg |
350.80€
100 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15886167
|
ACROBiosystems
CD3-H5257-1MG |
1 mg |
2153.18€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Myeloid cell surface antigen CD33 is also known as SIGLEC3, Siglecs (sialic acid binding Iglike lectins) and GP67, is a single-pass type I membrane protein which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Human CD33 / Siglec-3 cDNA encodes a 364 amino acid (aa) polypeptide with a hydrophobic signal peptide, an N-terminal Ig-like V-type domain, one Ig-like C2-type domains, a transmembrane region and a cytoplasmic tail. CD33 / Siglec-3 usually considered myeloid-specific, but it can also be found on some lymphoid cells. In the immune response, CD33 / Siglec-3 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. CD33 / Siglec-3 induces apoptosis in acute myeloid leukemia.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 64-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH28152.1 | |
Human Siglec-3, Fc Tag (CD3-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). | |
-20°C | |
Siglec-3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
52.9 kDa | |
Asp 18 - His 259 | |
CD33,SIGLEC3,gp67 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |