Learn More
Abnova™ Human SNX15 Partial ORF (NP_037438, 51 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00029907-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: RYSDFRKLHGDLAYTHRNLFRRLEEFPAFPRAQVFGRFEASVIEERRKGAEDLLRFTVHIPALNNSPQLKEFFRGGEVTRPLEVSRDLHEspecificaciones
NP_037438 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RYSDFRKLHGDLAYTHRNLFRRLEEFPAFPRAQVFGRFEASVIEERRKGAEDLLRFTVHIPALNNSPQLKEFFRGGEVTRPLEVSRDLH | |
RUO | |
SNX15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29907 | |
SNX15 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSAF001435 | |
SNX15 | |
Recombinant | |
wheat germ expression system |