Learn More
Abnova™ Human THSD3 Full-length ORF (NP_954874.1, 1 a.a. - 483 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00145501-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene contains a type 1 thrombospondin domain, which is present in thrombospondin, a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MTPGNATPPRTPEVTPLRLELQKLPGLANTTLSTPNPDTQASASPDPRPLREEEEARLLPRTHLQAELHQHGCWTVTEPAALTPGNATPPRTQEVTPLLLELQKLPELVHATLSTPNPDNQVTIKVVEDPQAEVSIDLLAEPSNPPPQDTLSWLPALWSFLWGDYKGEEKDRAPGEKGEEKEEDEDYPSEDIEGEDQEDKEEDEEEQALWFNGTTDNWDQGWLAPGDWVFKDSVSYDYEPQKEWSPWSPCSGNCSTGKQQRTRPCGYGCTATETRTCDLPSCPGTEDKDTLGLPSEEWKLLARNATDMHDQDVDSCEKWLNCKSDFLIKYLSQMLRDLPSCPCAYPLEAMDSPVSLQDEHQGRSFRWRDASGPRERLDIYQPTARFCLRSMLSGESSTLAAQHCCYDEDSRLLTRGKGAGMPNLISTDFSPKLHFKFDTTPWILCKGDWSRLHAVLPPNNGRACTDNPLEEEYLAQLQEAKEYEspecificaciones
NP_954874.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E0215/FLJ32147/MGC119416/TAIL1/THSD3 | |
ISM2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
145501 | |
THSD3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTPGNATPPRTPEVTPLRLELQKLPGLANTTLSTPNPDTQASASPDPRPLREEEEARLLPRTHLQAELHQHGCWTVTEPAALTPGNATPPRTQEVTPLLLELQKLPELVHATLSTPNPDNQVTIKVVEDPQAEVSIDLLAEPSNPPPQDTLSWLPALWSFLWGDYKGEEKDRAPGEKGEEKEEDEDYPSEDIEGEDQEDKEEDEEEQALWFNGTTDNWDQGWLAPGDWVFKDSVSYDYEPQKEWSPWSPCSGNCSTGKQQRTRPCGYGCTATETRTCDLPSCPGTEDKDTLGLPSEEWKLLARNATDMHDQDVDSCEKWLNCKSDFLIKYLSQMLRDLPSCPCAYPLEAMDSPVSLQDEHQGRSFRWRDASGPRERLDIYQPTARFCLRSMLSGESSTLAAQHCCYDEDSRLLTRGKGAGMPNLISTDFSPKLHFKFDTTPWILCKGDWSRLHAVLPPNNGRACTDNPLEEEYLAQLQEAKEY | |
RUO | |
ISM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |