missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TLE3 Full-length ORF (AAH41831.1, 1 a.a. - 772 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007090-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16171422

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 04-09-2024
para ver el stock

Descripción

Descripción

Sequence: MPPPPPLSCLRGLQAPHQPGQPGFKFTVAESCDRIKDEFQFLQAQYHSLKVEYDKLANEKTEMQRHYVMYYEMSYGLNIEMHKQTEIAKRLNTILAQIMPFLSQEHQQQVAQAVERAKQVTMTELNAIIGQQLQAQHLSHATHGPPVQLPPHPSGLQPPGIPPVTGSSSGLLALGALGSQAHLTVKDEKNHHELDHRERESSANNSVSPSESLRASEKHRGSADYSMEAKKRKAEEKDSLSRYDSDGDKSDDLVVDVSNEDPATPRVSPAHSPPENGLDKARSLKKDAPTSPASVASSSSTPSSKTKDLGHNDKSSTPGLKSNTPTPRNDAPTPGTSTTPGLRSMPGKPPGMDPIGIMASALRTPISITSSYAAPFAMMSHHEMNGSLTSPGAYAGLHNIPPQMSAAAAAAAATYGRSPMVGFDPHPPMRATGLPSSLASIPGGKPAYSFHVSADGQMQPVPFPHDALAGPGIPRHARQINTLSHGEVVCAVTISNPTRHVYTGGKGCVKIWDISQPGSKSPISQLDCLNRDNYIRSCKLLPDGRTLIVGGEASTLTIWDLASPTPRIKAELTSSAPACYALAISPDAKVCFSCCSDGNIAVWDLHNQTLVRQFQGHTDGASCIDISHDGTKLWTGGLDNTVRSWDLREGRQLQQHDFTSQIFSLGYCPTGEWLAVGMESSNVEVLHHTKPDKYQLHLHESCVLSLKFAYCGKWFVSTGKDNLLNAWRTPYGASIFQSKESSSVLSCDISADDKYIVTGSGDKKATVYEVIY
Especificaciones

Especificaciones

AAH41831.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ESG/ESG3/FLJ39460/GRG3/HsT18976/KIAA1547
TLE3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7090
TLE3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPPPPPLSCLRGLQAPHQPGQPGFKFTVAESCDRIKDEFQFLQAQYHSLKVEYDKLANEKTEMQRHYVMYYEMSYGLNIEMHKQTEIAKRLNTILAQIMPFLSQEHQQQVAQAVERAKQVTMTELNAIIGQQLQAQHLSHATHGPPVQLPPHPSGLQPPGIPPVTGSSSGLLALGALGSQAHLTVKDEKNHHELDHRERESSANNSVSPSESLRASEKHRGSADYSMEAKKRKAEEKDSLSRYDSDGDKSDDLVVDVSNEDPATPRVSPAHSPPENGLDKARSLKKDAPTSPASVASSSSTPSSKTKDLGHNDKSSTPGLKSNTPTPRNDAPTPGTSTTPGLRSMPGKPPGMDPIGIMASALRTPISITSSYAAPFAMMSHHEMNGSLTSPGAYAGLHNIPPQMSAAAAAAAATYGRSPMVGFDPHPPMRATGLPSSLASIPGGKPAYSFHVSADGQMQPVPFPHDALAGPGIPRHARQINTLSHGEVVCAVTISNPTRHVYTGGKGCVKIWDISQPGSKSPISQLDCLNRDNYIRSCKLLPDGRTLIVGGEASTLTIWDLASPTPRIKAELTSSAPACYALAISPDAKVCFSCCSDGNIAVWDLHNQTLVRQFQGHTDGASCIDISHDGTKLWTGGLDNTVRSWDLREGRQLQQHDFTSQIFSLGYCPTGEWLAVGMESSNVEVLHHTKPDKYQLHLHESCVLSLKFAYCGKWFVSTGKDNLLNAWRTPYGASIFQSKESSSVLSCDISADDKYIVTGSGDKKATVYEVIY
RUO
TLE3
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales