missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human uPAR / PLAUR Protein, His Tag
302.42€ - 1318.52€
Especificaciones
Número de acceso | AAH02788 |
---|---|
Peso molecular | 32.3 kDa |
Fuente | Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # AAH02788). |
Inmunógeno | Leu 23 - Arg 303 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15807897
|
ACROBiosystems
UPR-H5226-200UG |
200 μg |
302.42€
200 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15897887
|
ACROBiosystems
UPR-H5226-1MG |
1 mg |
1318.52€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Description
Urokinase plasminogen activator surface receptor (U-PAR) is also known as PLAUR, Monocyte activation antigen Mo3, CD antigen CD87. PLAUR contains three UPAR/Ly6 domains. U-PAR is expressed in neurons of the rolandic area of the brain (at protein level) and is also expressed in the brain. PLAUR / CD87 interacts with MRC2, SRPX2 and SORL1. PLAUR / UPAR acts as a receptor for urokinase plasminogen activator and plays a role in localizing and promoting plasmin formation. U-PAR mediates the proteolysis-independent signal transduction activation effects of U-PA.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.3 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH02788 | |
Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # AAH02788). | |
-20°C | |
uPAR | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
32.3 kDa | |
Leu 23 - Arg 303 | |
uPAR,PLAUR,CD87,MO3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |