missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human VEGF121 Protein, His Tag
411.28€ - 3362.84€
Especificaciones
Peso molecular | 14.9 kDa |
---|---|
Fuente | Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # P15692-9). |
Inmunógeno | Ala 27 - Arg 147 |
Requisitos de almacenamiento | -20°C |
Alias de gen | RP1-261G23.1,MGC70609,MVCD1,VEGFA,VPF |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15877907
|
ACROBiosystems
VE1-H5246-1MG |
1 mg |
3362.84€
1 miligramo |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15887907
|
ACROBiosystems
VE1-H5246-50UG |
50 μg |
411.28€
50 microgramos |
Fecha estimada de envío: 07-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Vascular endothelial growth factor (VEGF), also known as vascular permeability factor (VPF) and VEGF-A, and is a member of the platelet-derived growth factor (PDGF)/vascular endothelial growth factor (VEGF) family and encodes a protein that is often found as a disulfide linked homodimer. This protein is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. Alternatively spliced isoforms of 121,145,165,183,189 and 206 amino acids in length are expressed in humans. VEGF165 appears to be the most abundant and potent isoform, followed by VEGF121 and VEGF189. VEGF121 is the only form that lacks a basic heparinbinding region and is freely diffusible. Mouse embryos expressing only the corresponding isoform (VEGF120) do not survive to term, and show defects in skeletogenesis. Human VEGF121 shares 87% aa sequence identity with corresponding regions of mouse and rat, 93% with feline, equine and bovine, and 91%, 95% and 96% with ovine, canine and porcine VEGF, respectively. VEGF121 induces the proliferation of lymphatic endothelial cells. The lymphangiogenesis may be promoted by upregulation of VEGF121, which may in turn act in part via induction of VEGF-C. This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 14.9 kDa. As a result of glycosylation, the protein migrates as 17 kDa and 20 kDa under reducing (R) condition, and 35-45 kDa under non-reducing (NR) conditionEspecificaciones
14.9 kDa | |
Ala 27 - Arg 147 | |
RP1-261G23.1,MGC70609,MVCD1,VEGFA,VPF | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |
Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # P15692-9). | |
-20°C | |
VEGF121 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |