Learn More
Novus Biologicals™ Human Vein Whole Tissue Lysate (Adult Whole Normal)
Marca:  Novus Biologicals™ NB820-59176
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18218043
  • 508.63€ / 1 miligramo
Fecha estimada de envío: 28-08-2024
para ver el stock
Descripción
Descripción
The Human Vein Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Especificaciones
Especificaciones
Western Blot
1 mg
Total Protein - Human Adult Normal Tissue: Blood Vessel: Vein
Normal
Human
Human Vein Whole Tissue Lysate (Adult Normal)
Adult
Blood Lymph
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only