Learn More
Novus Biologicals™ Human Vein Whole Tissue Lysate (Adult Whole Normal)
Marca: Novus Biologicals™ NB820-59176
Detalles adicionales : Peso : 0.00970kg
Descripción
The Human Vein Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Especificaciones
Western Blot | |
1 mg | |
Total Protein - Human Adult Normal Tissue: Blood Vessel: Vein | |
Normal | |
Human | |
Human Vein Whole Tissue Lysate (Adult Normal) | |
Adult |
Blood Lymph | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only