Learn More
ACROBiosystems Human VLDL R Protein, His Tag
Marca: ACROBiosystems VLR-H5227-1MG
Detalles adicionales : Peso : 0.00001kg
Descripción
The very-low-density-lipoprotein receptor (VLDL-R) is a lipoprotein receptor that shows considerable similarity to the lowdensity-lipoprotein receptor. VLDL R is a 130 kDa type I transmembrane protein in the LDL receptor family that plays a significant role in lipid metabolism and in nervous system development and function .This receptor has been suggested to be important for the metabolism of apoprotein-E-containing triacylglycerol-rich lipoproteins, such as very-low-densitylipoprotein (VLDL), beta-migrating VLDL and intermediate-density lipoprotein. It is also one of the receptors of reelin, an extracellular matrix protein which regulates the processes of neuronal migration and synaptic plasticity. In humans, the VLDL-R is encoded by the VLDLR gene. A rare neurological disorder first described in the 1970s under the name “disequilibrium syndrome” is now considered to be caused by the disruption of VLDLR gene. The disorder was renamed VLDLR-associated cerebellar hypoplasia (VLDLRCH) after a 2005 study. It is associated with parental consanguinity and found in secluded communities such as the Hutterites. VLDLRCH is one of the two known genetic disorders caused by a disruption of reelin signaling pathway, along with Norman-Roberts syndrome.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 84.0 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Especificaciones
NP_001018066.1 | |
1 mg | |
Gly 28 - Ser 769 | |
VLDLR,RP11-320E16.1,CHRMQ1,FLJ35024,VLDLRCH,VLDL receptor | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
84.0 kDa | |
Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession # P98155-2). | |
-20°C | |
VLDL R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |