Learn More
R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit
Click to view available options
Cantidad:
10 plex
11 plex
12 plex
13 plex
14 plex
15 plex
16 plex
17 plex
18 plex
19 plex
2 plex
20 plex
21 plex
22 plex
23 plex
24 plex
25 plex
26 plex
27 plex
28 plex
29 plex
3 plex
30 plex
31 plex
32 plex
33 plex
34 plex
35 plex
36 plex
37 plex
38 plex
39 plex
4 plex
40 plex
41 plex
42 plex
43 plex
44 plex
45 plex
46 plex
5 plex
6 plex
7 plex
8 plex
9 plex
missing translation for 'unitSize'
cada uno
Specifications
Specifications
| Cantidad | 10 plex |
| Especie | Human |
| Para utilizar con (aplicación) | ELISA |
| Contenido y almacenamiento | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction