missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IA-6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA541650
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: KRTGGLYRVR LAERVFPLLG PQGAPPFLEE APSASLPGAE RATPPTREEP Sequence homology: Human: 100%
IA-6 Polyclonal antibody specifically detects IA-6 in Human samples. It is validated for Western Blot.Especificaciones
IA-6 | |
Polyclonal | |
Unconjugated | |
INSM2 | |
IA6; IA-6; INSM transcriptional repressor 2; Insm2; INSM2 variant 1; insulinoma-associated 2; insulinoma-associated protein 2; insulinoma-associated protein IA-6; methylated in liver tumor 1; mLt 1; mLt1; Nbla106; Zinc finger protein IA-6 | |
Rabbit | |
Protein A | |
RUO | |
84684 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96T92 | |
INSM2 | |
Synthetic peptide directed towards the N-terminal of human INSM2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |