Learn More
IFN gamma Recombinant Polyclonal Antibody (11HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marca: Invitrogen 710287
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with non-human primate, mouse and rat based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IFN gamma Recombinant Polyclonal antibody specifically detects IFN gamma in Human samples. It is validated for Flow Cytometry, Western Blot.Especificaciones
IFN gamma | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
Recombinant protein corresponding to amino acids 24–161 of human IFN-gamma | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Western Blot | |
11HCLC | |
Unconjugated | |
P01579 | |
Rabbit | |
Protein A | |
RUO | |
3458 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |