missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (gamma heavy chain) Rabbit anti-Human, Clone: RM116, Abnova™
Rabbit monoclonal antibody raised against native human IgG (gamma heavy chain).
Marca: Abnova MAB12808
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
IgG (gamma heavy chain) | |
Monoclonal | |
Unconjugated | |
ELISA (0.05-0.2 ug/mL for detection; 0.05-0.2 ug/well for capture) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
MGC117419 | |
Rabbit | |
Protein A affinity purification | |
RUO | |
3500/3501/3502/3503 | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
RM116 | |
Rabbit monoclonal antibody raised against native human IgG (gamma heavy chain). | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
IGHG1 | |
Native purified IgG from human plasma. | |
100 μg | |
Primary | |
This antibody reacts to the Fc region of all gamma heavy chains of human immunoglobulins, including gamma 1, gamma2, gamma 3, and gamma 4. No cross reactivity with other human heavy chains, mouse IgG, rat IgG or goat IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |