missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK gamma Monoclonal Antibody (H.256.9), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515155
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
It is not recommended to aliquot this antibody.
IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.Especificaciones
IKK gamma | |
Monoclonal | |
2.7 mg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
Q6TMG5, Q9Y6K9 | |
Ikbkg | |
Full length human GST-IKK gamma protein. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
H.256.9 | |
Unconjugated | |
Ikbkg | |
1110037D23Rik; AI848108; AI851264; AMCBX1; AW124339; FIP3; FIP-3; Fip3p; gikk; I-kappa-B kinase gamma; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; ikB kinase-associated protein 1; IKBKG; IKK[g]; IKK3; IKKAP1; IKKG; IKK-gamma; IMD33; incontinentia pigmenti; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma; inhibitor of kappaB kinase gamma; inhibitor of nuclear factor kappa-B kinase subunit gamma; IP; IP1; IP2; IPD2; mF; mFIP-3; Nemo; NF-kappa B essential modulator; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188; ZC2HC9 | |
Mouse | |
Affinity chromatography | |
RUO | |
309295, 8517 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido