Learn More
IL-1 alpha Monoclonal Antibody (4414), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA523694
Detalles adicionales : Peso : 0.00010kg
Descripción
In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-1 beta, -2, -3, -4, -5, -6, -7, -8, -9, -10, -11, -12, -13, rmIL-1 alpha, -1 beta, -3, -4, -5, -6, -7, -9, or -13. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-1 alpha Monoclonal antibody specifically detects IL-1 alpha in Human samples. It is validated for ELISA, Immunocytochemistry, Neutralization, Western Blot.Especificaciones
IL-1 alpha | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P01583 | |
Il1a | |
E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry, Neutralization, Western Blot | |
4414 | |
Unconjugated | |
Il1a | |
CTLA8; hematopoietin-1; IL 1 a; IL 1a; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1a; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 | |
Mouse | |
Protein A/G | |
RUO | |
3552 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |