Learn More
IL-10 Monoclonal Antibody (JES3-12G8), Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA182664
Detalles adicionales : Peso : 1.24740kg
Descripción
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for this product is human tonsil. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Human Interleukin-10 antibody, clone JES3-12G8 recognizes human Interleukin-10 (IL-10).
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Especificaciones
IL-10 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P22301 | |
IL10 | |
Human IL-10 expressed by COS cells. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
JES3-12G8 | |
Unconjugated | |
IL10 | |
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
Rat | |
Protein G | |
RUO | |
3586 | |
4° C, do not freeze | |
Liquid |