missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1RAcP Monoclonal Antibody (04), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543520
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human IL-1RAcP/IL-1R3
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.Especificaciones
IL-1RAcP | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NPH3 | |
IL1RAP | |
Recombinant Human IL-1RAcP/IL-1R3 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
04 | |
Unconjugated | |
IL1RAP | |
6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1R-3; IL1RAcP; IL-1RAcp; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain | |
Mouse | |
Protein A | |
RUO | |
3556 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |