missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL12RB1 Monoclonal Antibody (05), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA543495
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human IL12RB1
The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukin 12 with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Especificaciones
| IL12RB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P42701 | |
| IL12RB1 | |
| Recombinant Human IL12RB1 Protein. | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 05 | |
| Unconjugated | |
| IL12RB1 | |
| CD212; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL12R; IL-12R subunit beta-1; IL-12R[b]; IL12RB; Il12rb1; IL-12RB1; IL-12R-BETA1; IL-12R-beta-1; IMD30; interleukin 12 receptor subunit beta 1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor beta-1 chain precursor (IL-12R-beta1) (Interleukin-12 receptor beta) (IL-12 receptor beta component); interleukin-12 receptor subunit beta-1; MGC34454 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3594 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido