Learn More
IL17RA Monoclonal Antibody (133617), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA523864
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL&#x2011;17 R, rmIL-17B R, or recombinant human IL-17B R is observed. In Western blots, no cross-reactivity with rmIL-17 R is observed Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL17RA Monoclonal antibody specifically detects IL17RA in Human samples. It is validated for Flow Cytometry, Neutralization, Western Blot.Especificaciones
IL17RA | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96F46 | |
Il17ra | |
Mouse myeloma cell line NS0-derived recombinant human IL-17RA/IL-17R Leu33-Trp320 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Neutralization, Western Blot | |
133617 | |
Unconjugated | |
Il17ra | |
AW538159; CANDF5; CD127; CD217; CDw217; hIL-17R; IL-17 receptor A; IL17R; Il17ra; IL-17RA; interleukin 17 receptor A; interleukin-17 receptor A; MGC10262; VDw217 | |
Mouse | |
Protein A/G | |
RUO | |
23765 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |