Learn More
IL3RA Mouse anti-Human, Clone: 10B8E7, Abnova™
Mouse monoclonal antibody raised against partial recombinant human IL3RA.
Marca: Abnova MAB12487.50uL
Descripción
The protein encoded by this gene is an interleukin 3 specific subunit of a heterodimeric cytokine receptor. The receptor is comprised of a ligand specific alpha subunit and a signal transducing beta subunit shared by the receptors for interleukin 3 (IL3), colony stimulating factor 2 (CSF2/GM-CSF), and interleukin 5 (IL5). The binding of this protein to IL3 depends on the beta subunit. The beta subunit is activated by the ligand binding, and is required for the biological activities of IL3. This gene and the gene encoding the colony stimulating factor 2 receptor alpha chain (CSF2RA) form a cytokine receptor gene cluster in a X-Y pseudoautosomal region on chromosomes X or Y. [provided by RefSeq]
Especificaciones
IL3RA | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry Immunocytochemistry (1:200-1:100) Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determ | |
CD123/IL3R/IL3RAY/IL3RX/IL3RY/MGC34174/hIL-3Ra | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
10B8E7 | |
Mouse monoclonal antibody raised against partial recombinant human IL3RA. | |
In ascites (0.03% sodium azide) | |
IL3RA | |
Recombinant protein corresponding to amino acids 200-305 of human IL3RA. | |
RUO | |
3563 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.