Learn More
IMPACT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559805
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: ENKKIASATH NIYAYRIYCE DKQTFLQDCE DDGETAAGGR LLHLMEILNV KNVMVVV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Translational regulator that ensures constant high levels of translation upon a variety of stress conditions, such as amino acid starvation, UV-C irradiation, proteasome inhibitor treatment and glucose deprivation. Plays a role as a negative regulator of the EIF2AK4/GCN2 kinase activity; impairs GCN1-mediated EIF2AK4/GCN2 activation, and hence EIF2AK4/GCN2-mediated eIF-2-alpha phosphorylation and subsequent down-regulation of protein synthesis. May be required to regulate translation in specific neuronal cells under amino acid starvation conditions by preventing GCN2 activation and therefore ATF4 synthesis. Through its inhibitory action on EIF2AK4/GCN2, plays a role in differentiation of neuronal cells by stimulating neurite outgrowth.Especificaciones
IMPACT | |
Polyclonal | |
Unconjugated | |
IMPACT | |
E430016J11Rik; Impact; Impact homolog; impact RWD domain protein; impact, RWD domain protein; imprinted and ancient gene protein; Imprinted and ancient gene protein homolog; protein IMPACT; RWD domain containing 5; RWDD5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55364 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2X3 | |
IMPACT | |
Recombinant protein corresponding to Human IMPACT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only