Learn More
Tocris Bioscience™ Indocyanine green
Near-infrared fluorescent dye
Marca:  Tocris Bioscience™ 7510/50
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18738376
  • 193.30€ / 50 miligramos
Fecha estimada de envío: 05-08-2024
para ver el stock
Descripción
Descripción
Indocyanine green is a near infrared (NIR) fluorescent dye operating in the 700-900 nm wavelength range. It also exhibits fluorescence emission in the shortwave infrared window (SWIR; 1,000-2,000 nm) which enables in vivo imaging, including intravital microscopy, non-invasive real-time imaging in blood and lymph vessels, and imaging of hepatobiliary clearance. Imaging of indocyanine green in SWIR shows increased sensitivity, contrast, and resolution of fine anatomical structures over NIR.
This product is supplied with a high degree of hydration and may contain residual inorganic salts, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.
Especificaciones
Especificaciones
Sodium 4-(2-((1 E,3 E,5 E,7 Z)-7-(1,1-Dimethyl-3-(4-sulfonatobutyl)-1,3-dihydro-2 H-benzo[e]indol-2-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1 H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate
50 mg
C43H47N2O6S2Na
3599-32-4
Fluorescent Dyes
0.95