Learn More
INF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551447
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: GVARISDALL QLTCVSCVRA VMNSRQGIEY ILSNQGYVRQ LSQALDTSNV MVKKQVFELL AALCIYSPEG HVLTLDALDH YKTVCSQQYR FSIVMNELSG SDNVPYVVTL LSVINAVILG PEDLRARTQL RNEFIGLQLL DVLAR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
This gene represents a member of the formin family of proteins. It is considered a diaphanous formin due to the presence of a diaphanous inhibitory domain located at the N-terminus of the encoded protein. Studies of a similar mouse protein indicate that the protein encoded by this locus may function in polymerization and depolymerization of actin filaments. Mutations at this locus have been associated with focal segmental glomerulosclerosis 5.Especificaciones
INF2 | |
Polyclonal | |
Unconjugated | |
INF2 | |
2610204M08Rik; AA589465; AW125550; C14orf151; C14orf173; CMTDIE; EG629699; formin, inverted; FSGS5; HBEAG-binding protein 2 binding protein C; HBEBP2-binding protein C; Inf2; inverted formin; inverted formin 2; inverted formin, FH2 and WH2 domain containing; inverted formin-2; MGC13251; pp9484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64423 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q27J81 | |
INF2 | |
Recombinant protein corresponding to Human INF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only