Learn More
Influenza A Virus H1N1 Hemagglutinin Antibody, R&D Systems™
Descripción
Hemagglutinin/HA Peptide Monoclonal antibody specifically detects Hemagglutinin/HA Peptide in Influenza A virus H1N1 samples. It is validated for Western Blot, Simple Western, Immunocytochemistry
Especificaciones
Especificaciones
| Antígeno | Hemagglutinin/HA Peptide |
| Aplicaciones | Western Blot, Simple Western, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 2908F |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1 μg/mL, Simple Western 25 μg/mL, Immunocytochemistry 3-15 μg/mL |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| N.º de referencia del gen | YP_009118626 |
| Especie del huésped | Rabbit |
| Inmunógeno | Human embryonic kidney cell, HEK293-derived influenza a virus H1N1 Hemagglutinin with a C-terminal 6-His tag, Met1-Gln529, Accession # YP_009118626 |
| Mostrar más |