missing translation for 'onlineSavingsMsg'
Learn More

ING4 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 67754-1-IG-20UL

 Ver más versiones de este producto

Código de producto. 30236412

  • ONLINE EXCLUSIVE PRICE
    145.00€
    129.00€ / 20 microlitros
    Ahorro 16.00€
    11% Descuento

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

The ING4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ING4. This antibody recognizes human, pig, rabbit, rat antigen. The ING4 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.
Especificaciones

Especificaciones

ING4
Monoclonal
1000 μg/mL
MGC12557; my036; p29ING4
Recombinant Protein
20 μL
Primary
Human, Pig, Rabbit, Rat
29 kDa
Liquid
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1A12A3
Unconjugated
Mouse
Protein G purification
RUO
51147
Store at -20°C. Stable for one year after shipment.
Monoclonal Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.