missing translation for 'onlineSavingsMsg'
Learn More

Insulin/Proinsulin Antibody Cocktail, Invitrogen™

Antibody Cocktail

Brand:  Invitrogen MA512042

Product Code. 11354563

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products »

This item is not returnable. View return policy

Description

Description

MA5-12042 targets Insulin (INS04 + INS05) in IHC (P) applications and shows reactivity with Human, Porcine, and Rat samples. The MA5-12042 immunogen is insulin protein.

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insulin/Proinsulin
Cocktail
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P01308, P01315, P01322, P01323
INS, Ins1, Ins2
Insulin protein
500 μL
Primary
Human, Pig, Rat
Antibody
IgG1 κ
Immunohistochemistry (Paraffin)
INS04, INS05
Unconjugated
INS
AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin
Mouse
Protein G
RUO
24505, 24506, 3630, 397415
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.