Learn More
Integrin beta 7 Monoclonal Antibody (473207), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Thermo Scientific MA523592
Detalles adicionales : Peso : 0.00010kg
Descripción
In Western blots, no cross-reactivity with recombinant human Integrin beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 8, recombinant mouse Integrin beta 4, or beta 7 is observed.
Integrin beta 7 Monoclonal antibody specifically detects Integrin beta 7 in Human samples. It is validated for Flow Cytometry.Especificaciones
Integrin beta 7 | |
Monoclonal | |
FITC | |
ITGB7 | |
Gut homing receptor beta subunit; integrin beta 7; integrin beta 7 subunit; integrin beta-7; Integrin beta-P; integrin subunit beta 7; integrin, beta 7; ITGB7; Ly69; M290 IEL antigen | |
Mouse | |
Protein A/G | |
RUO | |
3695 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
473207 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P26010 | |
ITGB7 | |
CHO-derived recombinant human Integrin beta 7 long isoform Glu20-His723 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |