Learn More
Tuttnauer Isothermal/Disinfection (for Agar Preparation)
Tienda Tuttnauer Productos
Click to view available options
:
Isothermal/Disinfection; Application: Agar Preparation
Descripción
- Disinfection (agar prep)
- Isothermal/Disinfection (for Agar preparation).
- The isothermal cycle has a temperature range from 60°C to 94°C and allows for gentle heating and cooling of agar.
Especificaciones
Especificaciones
| Para utilizar con (aplicación) | Agar Preparation |
| Tipo de producto | Software |