Learn More
ITGA7 Monoclonal Antibody (334908), PE, Invitrogen™
Rat Monoclonal Antibody
Marca: Thermo Scientific MA523608
Detalles adicionales : Peso : 0.00010kg
Descripción
Reactivity with other Integrin alpha 7 isoforms was not tested. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 2, alpha 3, alpha 4, alpha 5, alpha 9, alpha E, alpha X, recombinant human Integrin alpha L, or alpha 11 is observed.
ITGA7 Monoclonal antibody specifically detects ITGA7 in Mouse samples. It is validated for Flow Cytometry.Especificaciones
ITGA7 | |
Monoclonal | |
PE | |
Itga7 | |
[a]7; alpha 7A integrin; alpha7; H36-alpha7; integrin alpha 7; integrin alpha 7 chain; integrin alpha-7; Integrin alpha-7 70 kDa form; Integrin alpha-7 heavy chain; Integrin alpha-7 light chain; integrin subunit alpha 7; integrin, alpha 7; ITGA7; UNQ406/PRO768 | |
Rat | |
Protein A/G | |
RUO | |
16404 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
334908 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q61738 | |
Itga7 | |
CHO-derived recombinant mouse Integrin alpha 7 Phe34-Pro1036 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2b |