Learn More
ITGA9 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA547540
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human Integrin alpha 1, alpha 8, alpha 9, alpha L, recombinant mouse Integrin 3, 4, 5, E, and M is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ITGA9 Polyclonal antibody specifically detects ITGA9 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Western Blot.Especificaciones
ITGA9 | |
Polyclonal | |
Unconjugated | |
Itga9 | |
(alpha)9; 2610002H11Rik; 6720458D17Rik; AI461869; ALPHA-RLC; D130073C02; D9Ertd428e; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin subunit alpha 9; integrin, alpha 9; ITGA4L; Itga9; Itga9l; RLC | |
Goat | |
Antigen affinity chromatography | |
RUO | |
104099 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
B8JK39 | |
Itga9 | |
Mouse myeloma cell line NS0-derived recombinant mouse Integin alpha 9 Try31-Val979 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |