missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Japanese Encephalitis Virus NS4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532211
Detalles adicionales : Peso : 0.01000kg
Description
Recommended positive controls: BHK21 infected JEV. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Japanese encephalitis virus (JEV) is mosquito born flavivirus that induces acute encephalitis in tropical and subtropical world.Specifications
Japanese Encephalitis Virus NS4B | |
Polyclonal | |
Unconjugated | |
Flavivirus; JEV; NS4B | |
Full length Japanese encephalitis virus JEV NS4b Recombinant protein (Japanese Encephalitis Virus strain Jaoars982) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.76 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |