Learn More
Invitrogen™ JunD Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA1834
Descripción
PA1-834 detects JunD from mouse cells. PA1-834 has been successfully used in Western blot and ChIP procedures. By Western blot, this antibody detects proteins from ~37-41 kDa representing JunD from both un-induced and induced mouse NIH 3T3 cells. The PA1-834 immunogen is a synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD. This sequence is comletely conserved between mouse, human and rat. Reconstitute with PBS.
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
Especificaciones
| JunD | |
| Polyclonal | |
| Unconjugated | |
| JUND | |
| activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16478, 24518, 3727 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 3.04 mg/mL | |
| PBS with 0.05% sodium azide | |
| P15066, P17535, P52909 | |
| JUND | |
| Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.