Learn More
KappaB ras2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529432
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Atypical Ras-like protein that acts as a potent regulator of NF-kappa-B activity by preventing the degradation of NF-kappa-B inhibitor beta (NFKBIB) by most signals, explaining why NFKBIB is more resistant to degradation. May act by blocking phosphorylation of NFKBIB and nuclear localization of p65/RELA NF-kappa-B subunit. It is unclear whether it acts as a GTPase. Both GTP- and GDP-bound forms block phosphorylation of NFKBIB.Especificaciones
KappaB ras2 | |
Polyclonal | |
Unconjugated | |
NKIRAS2 | |
2410003M04Rik; 4930527H08Rik; D630018G21Rik; I-kappa-B-interacting Ras-like protein 2; Kappa B-Ras protein 2; kappaB-Ras2; KBRAS2; NF-kappa-B inhibitor-interacting Ras-like protein 2; NFKB inhibitor interacting Ras like 2; NFKB inhibitor interacting Ras-like 2; NFKB inhibitor interacting Ras-like protein 2; NKIRAS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9NYR9 | |
NKIRAS2 | |
Synthetic peptide corresponding to a region within amino acids 129 and 191 of Human NKIRAS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |