missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDR Mouse anti-Human, Clone: OTI2A1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF802443
Detalles adicionales : Peso : 0.05000kg
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.. Mutations of this gene are implicated in infantile capillary hemangiomas.Especificaciones
KDR | |
Monoclonal | |
Unconjugated | |
KDR | |
CD309, FLK1, VEGFR, VEGFR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3791 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P35968 | |
KDR | |
Human recombit protein fragment corresponding to amino acids 786-1356 of human KDR produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |