missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ki-67 Monoclonal Antibody (7B11), FITC

Descripción
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with euchromatin at the perichromosomal layer, the centromeric heterochromatin, and the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis. Ki-67 antibodies are useful in establishing the cell growing fraction in neoplasms. In neoplastic tissues, the prognostic value is comparable to the tritiated thymidine-labelling index. The correlation between low Ki-67 index and histologically low-grade tumors is strong. Ki-67 is routinely used as a neuronal marker of cell cycling and proliferation.
Especificaciones
Especificaciones
| Antígeno | Ki-67 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 7B11 |
| Concentración | 0.25mg/mL |
| Conjugado | FITC |
| Descripción | Ki-67 Monoclonal Antibody, 0.5mL, 0.25mg/mL Concentration, 7B11 Clone, Ki-67 Antigen, Ki-67 Synthetic Peptide Immunogen, Mouse Host, Human Species Reactivity, FITC Conjugate, IgG1 kappa Isotype, PBS with 4mg/mL BSA Storage Buffer, Used For Flow Cytometry |
| Dilución | Assay-dependent |
| Formulación | PBS with 4mg/mL BSA and 0.1% sodium azide |
| génica | Mki67 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?