Learn More
KISS1R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533877
369.35 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Bovine, Dog, Bat, Hamster, Pig (100%) Goat, Elephant, Panda (94%).
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.
Especificaciones
KISS1R | |
Polyclonal | |
Unconjugated | |
KISS1R | |
AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100753032, 114229, 612374, 733704, 78976, 84634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q91V45, Q924U1, Q969F8 | |
KISS1R | |
Synthetic 17 amino acid peptide from 2nd extracellular domain of human KISS1R / GPR54. | |
50 μg | |
Primary | |
Canine, Hamster, Human, Mouse, Non-human Primate, Pig, Rat | |
Antibody | |
IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.