missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLRG1 Monoclonal Antibody (2F1), Brilliant Ultra Violet™ 563, eBioscience™, Invitrogen™

Descripción
KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
Especificaciones
Especificaciones
| Antígeno | KLRG1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 2F1 |
| Concentración | 0.2 mg/mL |
| Conjugado | Brilliant Ultraviolet 563 |
| Formulación | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| génica | Klrg1 |
| N.º de referencia del gen | O88713 |
| Alias de gen | 2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?