missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT18 Mouse anti-Human, Clone: OTI1H10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF500017

Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.Especificaciones
KRT18 | |
Monoclonal | |
Unconjugated | |
KRT18 | |
CK-18, CYK18, K18 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3875 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P05783 | |
KRT18 | |
Human recombit protein fragment corresponding to amino acids 69-372 of human CK18 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
KRT18 Mouse anti-Human, Clone: OTI1H10, lyophilized, TrueMAB™
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido