Learn More
Thermo Scientific Chemicals Clorhidrato de dimetiléster del ácido L-aspártico, 98 %
Catalog Number p-7043171
Click to view available options
Cantidad:
5 g
25 g
32213-95-9
C6H12ClNO4
197.615
MFCD00038878
PNLXWGDXZOYUKB-WCCKRBBISA-N
h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
2734892
dimetil (2S)-2-aminobutanodioato;clorhidrato
COC(=O)CC(C(=O)OC)N.Cl
32213-95-9
C6H12ClNO4
197.615
MFCD00038878
PNLXWGDXZOYUKB-WCCKRBBISA-N
h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
2734892
dimetil (2S)-2-aminobutanodioato;clorhidrato
COC(=O)CC(C(=O)OC)N.Cl
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Es una importante materia prima e intermediario utilizado en la síntesis orgánica, los productos farmacéuticos, los agroquímicos y los colorantes.
Solubilidad
Soluble en agua (casi transparencia) y metanol.
Notas
Almacenar en fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material L-Aspartic acid dimethyl ester hydrochloride
Punto de fusión 115°C to 120°C
Color Blanco
Información de solubilidad Soluble in water (almost transparency) and methanol.
Formula Weight (peso de la fórmula) 197.62
Porcentaje de pureza 98%
Cantidad 5 g
Rotación óptica +15° (c=1 in Methanol)
Forma física Polvo