missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Calretinin Rabbit Monoclonal Antibody
Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ Calretinin, Rabbit Monoclonal Antibody.
Marca: Epredia™ RM-9113-R7
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.Host Species: Rabbit
Clone: SP13
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Not determined
Immunogen: Recombinant full-length mouse calretinin protein
Molecular Weight: 31.5 kDa
Positive Control: Mesothelioma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Calretinin | |
Monoclonal | |
Unconjugated | |
Recombinant full-length mouse calretinin protein | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
SP13 | |
Rabbit | |
7 mL | |
Oncogenesis | |
Human, Mouse | |
IgG |