missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Follicle Stimulating Hormone (FSH) Ab-3, Mouse Monoclonal Antibody, Epredia™
Epredia™ Follicle Stimulating Hormone (FSH) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.
Especificaciones
| Antígeno | Follicle Stimulating Hormone (FSH) Ab-3 |
|---|---|
| Clon | FSH03 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Conjugado | Unconjugated |
Descripción
FSH is a pituitary hormone involved in the maturation of ovarian follicles and estrogen secretion in females. In the pituitary gland, FSH is produced by gonadotrophs. In males, FSH stimulates the secretion of testosterone.Host Species: Mouse
Clone: FSH03
Isotype: IgG1/κ
Species Reactivity: Human and Cow. Others not known.
Immunogen: Purified human FSH-β
Positive Control: Anterior pituitary
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
| Follicle Stimulating Hormone (FSH) Ab-3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| IgG2a κ |
| FSH03 | |
| Monoclonal | |
| Mouse | |
| Purified human FSH-Beta |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto