Learn More
Ki-67 Ab-4 Rabbit Polyclonal Antibody, Epredia™
Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67 Ab-4, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.
Marca: Epredia RB-1510-P
Descripción
Ki67 is a nuclear protein, which is expressed in the proliferating cells. Ki67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiscent) phase are negative for this protein.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not known.
Epitope: Middle of Ki-67 protein
Immunogen: A synthetic peptide from the human Ki-67 protein
Positive Control: Tonsil or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Ki-67 Ab-4 | |
Polyclonal | |
1mg/mL | |
Rabbit | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Unconjugated | |
A synthetic peptide from the human Ki67 protein | |
RUO | |
Primary | |
Concentrated |