Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, two ports, Luer locks
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
533.00€
Especificaciones
Esterilizables en autoclave | Not Autoclavable |
---|---|
Certificaciones/conformidad | ISO 7 |
Color | Transparente |
Desechables | Single-use |
Para utilizar con (aplicación) | Bioprocessing |
Código de producto | Marca | Capacidad (métrico) | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Capacidad (métrico) | Precio | Cantidad y disponibilidad | |||||
17885716
|
Thermo Scientific™
SH30961.16 |
2 L |
533.00€
paquete de 10 |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Especificaciones
Not Autoclavable | |
Transparente | |
Bioprocessing | |
C-Flex | |
2 ports |
ISO 7 | |
Single-use | |
Estéril | |
Aegis 5-14 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.