missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Monoclonal Antibody (119CT80.1.1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA518107
346.50 EUR hasta el 2025-03-16
Use el código promocional "24070" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Lactoferrin (LF) is an iron binding glycoprotein. LF consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). LF is found in external fluids, including milk and mucosal secretions, and is a prominent component of secondary granuals of neutrophils. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. LTF acts as a major first line defense against microbial infections, partly due to its ability to efficiently bind to and hence remove Fe3+ from the environment, but studies have also shown that LTF possesses bactericidal properties and proteolytic activity, capable of cleaving arginine-rich sequences within microbial proteins. Three isoforms of LTF have been identified: LTF-alpha, LTF-beta and LTF-gamma and receptors for LTF are expressed in the gastrointestinal tract, by macrophages, lymphocytes, neutrophils and platelets and also by some bacteria.
Especificaciones
Lactoferrin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P02788 | |
LTF | |
LTF recombinant protein | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
119CT80.1.1 | |
Unconjugated | |
LTF | |
Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin | |
Mouse | |
Protein G | |
RUO | |
4057 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Lactoferrin Monoclonal Antibody (119CT80.1.1), Invitrogen™ > 400 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido