Learn More
Lamin B1 Antibody (919022), Unconjugated - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals MAB85251
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Lamin B1 Monoclonal antibody specifically detects Lamin B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
| Lamin B1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1 - 2 μg/mL | |
| ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419 | |
| E. coli-derived partial recombinant human Lamin B1 protein (amino acids 471-582) | |
| 100 μg | |
| Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors | |
| 4001 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 919022 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |