missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (508921), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB4800A
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LAMP-1/CD107a Monoclonal antibody specifically detects LAMP-1/CD107a in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Especificaciones
| LAMP-1/CD107a | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 508921 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| Mouse myeloma cell line NS0-derived recombinant human LAMP1/CD107a, Ala28-Asn380, Accession # P11279 | |
| 0.1 mL | |
| Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
| 3916 | |
| Store at 4C in the dark. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido