missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP1 Monoclonal Antibody (H4A3), PE, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen A15798
Detalles adicionales : Peso : 0.00280kg
Descripción
LAMP-1/CD107a Monoclonal antibody A15798 from Invitrogen specifically detects LAMP-1/CD107a in Human samples. It is validated for Flow Cytometry.
LAMP1 Monoclonal antibody specifically detects LAMP1 in Human, Mouse, Non-human Primate samples. It is validated for Flow Cytometry.Especificaciones
LAMP1 | |
Monoclonal | |
PE | |
LAMP1 | |
120 kDa lysosomal membrane glycoprotein; AI196048; CD107 antigen-like family member A; CD107a; I79_011073; Lamp I; LAMP1; LAMP-1; LAMPA; LGP120; LGP-120; LGPA; LGP-A; lysosomal associated membrane protein 1; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; lysosomal membrane glycoprotein A; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
16783, 3916 | |
4° C | |
IgG1 κ |
Flow Cytometry | |
H4A3 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P11279, P11438 | |
LAMP1 | |
Human adherent peripheral blood cells. | |
100 Tests | |
Primary | |
Human, Mouse, Non-human Primate | |
Liquid |