missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDHA Mouse anti-Human, Clone: OTI2D11, TrueMAB™, Origene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA500568S
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LDHA Monoclonal antibody specifically detects LDHA in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotEspecificaciones
LDHA | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P00338 | |
Mouse | |
Affinity chromatography | |
RUO | |
3939 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI2D11 | |
Unconjugated | |
LDHA | |
Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; GSD11; HEL-S-133P; l7R2; Lactate Dehydrogenase; lactate dehydrogenase 1, A chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase A; lactate dehydrogenase A4; lactate dehydrogenase A-like; lactate dehydrogenase C; lactate dehydrogenase C variant 3; lactate dehydrogenase M; lactate dehydrogenase-A; lactate dehydrogenase-M; lactate dehydrogenase-M (EC 1.1.1.27); Lactic Dehydrogenase; LDH; LDH muscle subunit; LDH testis subunit; Ldh1; Ldh-1; Ldh3; Ldha; LDH-A; ldha protein; ldha1; LDH-A4 muscle-specific isozyme; glycolytic enzyme; Ldhc; LDH-C; Ldhm; LDH-M; LDH-X; L-lactate dehydrogenase; L-lactate dehydrogenase A chain; L-lactate dehydrogenase C chain; M-LDH; OTTHUMP00000165206; PIG19; proliferation-inducing gene 19; renal carcinoma antigen NY-REN-59 | |
Full length human recombinant protein of human LDHA produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |