missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA536461
Detalles adicionales : Peso : 0.00910kg
Descripción
This antibody detects endogenous protein at a molecular weight of 53 kDa. Purity is >95% by SDS-PAGE.
LILRA1 Polyclonal antibody specifically detects LILRA1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry.Especificaciones
LILRA1 | |
Polyclonal | |
Unconjugated | |
LILRA1 | |
CD85 antigen-like family member I; CD85I; leucocyte Ig-like receptor A1; leukocyte immunoglobulin like receptor A1; leukocyte immunoglobulin-like receptor 6; Leukocyte immunoglobulin-like receptor subfamily A member 1; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1; LILRA1; LIR6; LIR-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11024 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O75019 | |
LILRA1 | |
Synthetic peptide corresponding to amino acids 51-80 of human LIR-6 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |