Learn More
LITAF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA522004
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (85%), Rat (86%), Rhesus Monkey (96%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene.Especificaciones
LITAF | |
Polyclonal | |
Unconjugated | |
LITAF | |
3222402J11Rik; C85531; CMT1C; EET-1; Estrogen-enhanced transcript protein; estrogen-enhanced transcript protein 1; estrogen-responsive uterine transcript; FLJ38636; lipopolysaccharide induced TNF factor; lipopolysaccharide-induced TNF factor; lipopolysaccharide-induced TNF-alpha factor; lipopolysaccharide-induced tumor necrosis factor-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor homolog; Litaf; LITAF-like protein; LPS-induced TN factor; LPS-induced TNF-alpha factor; LPS-induced TNF-alpha factor homolog; mEET; MGC116698; MGC116700; MGC116701; MGC125274; MGC125275; MGC125276; N4wbp3; Nedd4 WW domain-binding protein 3; p53-induced gene 7 protein; PIG 7; PIG7; SIMPLE; Small integral membrane protein of lysosome/late endosome; Tbx1; TP53I7; tumor protein p53 inducible protein 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9516 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q99732 | |
LITAF | |
Recombinant fragment corresponding to a region within amino acids 1 and 161 of LITAF (Uniprot ID#Q99732) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |